siRNA Details
| Gene Symbol: ECSCR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077693.3 | NP_001071161.1 | 6, 7 | 453 | ||
| NM_001293739.1 | NP_001280668.1 | 6, 7 | 453 | |||
| GenBank mRNA | BC146912.1 | - | 6, 7 | 425 | 9056840 | |
| BC146913.1 | - | 6, 7 | 425 | 9056841 | ||
| BI834795.1 | - | 6, 7 | 403 | |||
| DQ462572.1 | - | 6, 7 | 447 | |||
| EU025066.1 | - | 6, 7 | 453 | |||

ECSCR
Genomic Map