siRNA Details
| Gene Symbol: DPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129758.1 | NP_001123230.1 | 2 | 481 | ||
| NM_022357.3 | NP_071752.3 | 2 | 481 | |||
| GenBank mRNA | AJ291679.1 | - | 2 | 395 | ||
| AK057401.1 | - | 2 | 481 | |||
| AY358390.1 | - | 2 | 404 | |||
| BC037243.1 | - | 2 | 393 | |||
| BC057789.1 | - | 2 | 422 | 5297958 | ||
| DB020973.1 | - | 2 | 444 | |||

DPEP3
Genomic Map