siRNA Details
| Gene Symbol: MROH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100878.1 | NP_001094348.1 | 12, 13 | 1910 | ||
| XM_011517214.1 | XP_011515516.1 | 11, 12 | 2160 | |||
| XM_011517215.1 | XP_011515517.1 | 11, 12 | 2157 | |||
| XM_011517216.2 | XP_011515518.1 | 11, 12 | 2160 | |||
| XM_011517218.1 | XP_011515520.1 | 11, 12 | 1787 | |||
| XM_011517221.2 | XP_011515523.1 | 12, 13 | 1925 | |||
| XM_011517222.1 | XP_011515524.1 | 12, 13 | 1925 | |||
| GenBank mRNA | AF289596.1 | - | 1, 2 | 972 | ||
| AK292180.1 | - | 3, 4 | 558 | |||
| BC029031.1 | - | 3, 4 | 580 | |||

MROH6
Genomic Map