siRNA Details
Gene Symbol: KCNQ5-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120503.1 | 2 | 204 | |||
GenBank mRNA | BX104605.1 | - | 2 | 204 | ||
HY037383.1 | - | 2 | 345 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: KCNQ5-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120503.1 | 2 | 204 | |||
GenBank mRNA | BX104605.1 | - | 2 | 204 | ||
HY037383.1 | - | 2 | 345 |