siRNA Details
| Gene Symbol: KCNQ5-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120503.1 | 3 | 324 | |||
| GenBank mRNA | BX104605.1 | - | 3 | 324 | ||
| HY037383.1 | - | 3 | 465 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KCNQ5-IT1| Gene Symbol: KCNQ5-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120503.1 | 3 | 324 | |||
| GenBank mRNA | BX104605.1 | - | 3 | 324 | ||
| HY037383.1 | - | 3 | 465 | |||