siRNA Details
| Gene Symbol: ZNF487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026693.1 | 4 | 719 | |||
| GenBank mRNA | AK302442.1 | - | 4 | 719 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF487| Gene Symbol: ZNF487 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026693.1 | 4 | 719 | |||
| GenBank mRNA | AK302442.1 | - | 4 | 719 | ||