siRNA Details
| Gene Symbol: RAB17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022449.3 | NP_071894.1 | 5 | 1124 | ||
| NR_033308.1 | 4 | 972 | ||||
| XM_006712689.1 | XP_006712752.1 | 5 | 1007 | |||
| GenBank mRNA | AK022600.1 | - | 5 | 1107 | ||
| AK225423.1 | - | 5 | 1073 | |||
| BC000929.2 | - | 2 | 157 | |||
| BC007907.2 | - | 4 | 334 | |||
| BC009827.2 | - | 4 | 565 | |||
| BC050426.1 | - | 5 | 722 | 5761178 | ||
| BX647412.1 | - | 2 | 2346 | |||
| CR457282.1 | - | 4 | 461 | |||

RAB17
Genomic Map