siRNA Details
| Gene Symbol: FAM196A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039762.2 | NP_001034851.1 | 5 | 1790 | ||
| XM_005252694.3 | XP_005252751.1 | 3 | 1765 | |||
| XM_017016537.1 | XP_016872026.1 | 3 | 5361 | |||
| XM_017016538.1 | XP_016872027.1 | 4 | 4777 | |||
| XM_017016539.1 | XP_016872028.1 | 3 | 2504 | |||
| XM_017016540.1 | XP_016872029.1 | 4 | 2630 | |||
| XM_017016541.1 | XP_016872030.1 | 3 | 2066 | |||
| XM_017016542.1 | XP_016872031.1 | 4 | 4885 | |||
| GenBank mRNA | AK127465.1 | - | 5 | 1790 | ||
| BC131825.1 | - | 2 | 1275 | 40128007 | ||
| BC137112.1 | - | 3 | 1372 | 9021109 | ||

FAM196A
Genomic Map