siRNA Details
Gene Symbol: LOC643201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036494.1 | 7 | 915 | |||
GenBank mRNA | BC034407.1 | - | 7 | 980 | ||
BC052945.1 | - | 4 | 259 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC643201 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036494.1 | 7 | 915 | |||
GenBank mRNA | BC034407.1 | - | 7 | 980 | ||
BC052945.1 | - | 4 | 259 |