siRNA Details
| Gene Symbol: LOC643201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036494.1 | 10 | 1074 | |||
| GenBank mRNA | BC034407.1 | - | 10 | 1139 | ||
| BC052945.1 | - | 7 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643201| Gene Symbol: LOC643201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036494.1 | 10 | 1074 | |||
| GenBank mRNA | BC034407.1 | - | 10 | 1139 | ||
| BC052945.1 | - | 7 | 418 | |||