siRNA Details
| Gene Symbol: LOC643355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270945.1 | NP_001257874.1 | 1 | 222 | ||
| XM_005271117.3 | XP_005271174.1 | 2 | 311 | |||
| GenBank mRNA | BC034233.1 | - | 1 | 254 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC643355| Gene Symbol: LOC643355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270945.1 | NP_001257874.1 | 1 | 222 | ||
| XM_005271117.3 | XP_005271174.1 | 2 | 311 | |||
| GenBank mRNA | BC034233.1 | - | 1 | 254 | ||