siRNA Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290030.1 | NP_001276959.1 | 1, 2 | 507 | ||
| NM_001290031.1 | NP_001276960.1 | 1, 2 | 507 | |||
| XM_011510998.2 | XP_011509300.1 | 3, 4 | 639 | |||
| XM_011510999.2 | XP_011509301.1 | 2, 3 | 306 | |||
| XM_017003875.1 | XP_016859364.1 | 2, 3 | 1008 | |||
| XM_017003877.1 | XP_016859366.1 | 2, 3 | 306 | |||
| GenBank mRNA | AA401652.1 | - | 1, 2 | 58 | ||

ERICH2
Genomic Map