siRNA Details
| Gene Symbol: KRTAP27-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077711.1 | NP_001071179.1 | 1 | 428 | ||
| GenBank mRNA | AB096937.1 | - | 1 | 428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP27-1| Gene Symbol: KRTAP27-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077711.1 | NP_001071179.1 | 1 | 428 | ||
| GenBank mRNA | AB096937.1 | - | 1 | 428 | ||