siRNA Details
| Gene Symbol: CBLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039771.2 | NP_001034860.1 | 3 | 1065 | ||
| GenBank mRNA | AY359070.1 | - | 3 | 1065 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBLN3| Gene Symbol: CBLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039771.2 | NP_001034860.1 | 3 | 1065 | ||
| GenBank mRNA | AY359070.1 | - | 3 | 1065 | ||