siRNA Details
| Gene Symbol: CXCR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001716.4 | NP_001707.1 | 2 | 801 | ||
| NM_032966.2 | NP_116743.1 | 1 | 778 | |||
| GenBank mRNA | AK291512.1 | - | 2 | 801 | ||
| BC110352.1 | - | 2 | 715 | 5219901 | ||
| CO958128.1 | - | 1 | 510 | |||
| X68149.1 | - | 2 | 709 | |||
| X68829.1 | - | 1 | 778 | |||

CXCR5
Genomic Map