siRNA Details
| Gene Symbol: CDH24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022478.3 | NP_071923.2 | 7 | 1292 | ||
| NM_144985.3 | NP_659422.2 | 7 | 1292 | |||
| XM_011537089.1 | XP_011535391.1 | 7 | 1373 | |||
| GenBank mRNA | AK057922.1 | - | 7 | 1228 | ||
| AK289576.1 | - | 7 | 1292 | |||
| AK307746.1 | - | 7 | 1273 | |||
| AY260900.1 | - | 6 | 1125 | |||
| AY260901.1 | - | 6 | 1125 | |||
| AY358199.1 | - | 7 | 1248 | |||
| BX248303.1 | - | 3 | 316 | |||

CDH24
Genomic Map