siRNA Details
| Gene Symbol: GSG1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310219.1 | NP_001297148.1 | 3 | 393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GSG1L2| Gene Symbol: GSG1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310219.1 | NP_001297148.1 | 3 | 393 | ||