siRNA Details
| Gene Symbol: HCG2040054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120602.1 | 2, 3 | 343 | |||
| GenBank mRNA | AI150334.1 | - | 1, 2 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HCG2040054| Gene Symbol: HCG2040054 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120602.1 | 2, 3 | 343 | |||
| GenBank mRNA | AI150334.1 | - | 1, 2 | 90 | ||