siRNA Details
Gene Symbol: ARL14EPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195581.1 | NP_001182510.1 | 2 | 151 | ||
XM_017009722.1 | XP_016865211.1 | 3 | 245 | |||
GenBank mRNA | DT932734.1 | - | 214 |
