siRNA Details
| Gene Symbol: ARL14EPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195581.1 | NP_001182510.1 | 1, 2 | 105 | ||
| XM_017009722.1 | XP_016865211.1 | 2, 3 | 199 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARL14EPL| Gene Symbol: ARL14EPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195581.1 | NP_001182510.1 | 1, 2 | 105 | ||
| XM_017009722.1 | XP_016865211.1 | 2, 3 | 199 | |||