siRNA Details
| Gene Symbol: DNAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172810.1 | NP_001166281.1 | 6 | 734 | ||
| NM_023036.4 | NP_075462.3 | 6 | 737 | |||
| XM_011525125.2 | XP_011523427.1 | 6 | 754 | |||
| GenBank mRNA | AF250288.1 | - | 6 | 694 | ||
| AJ295276.1 | - | 5 | 933 | |||
| BC033013.1 | - | 5 | 611 | 4826432 | ||
| BC039582.1 | - | 6 | 724 | 5752153 | ||
| BG723530.1 | - | 5 | 611 | |||

DNAI2
Genomic Map