siRNA Details
| Gene Symbol: C15orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039905.2 | NP_001034994.1 | 1, 2 | 257 | ||
| GenBank mRNA | AK095915.1 | - | 1, 2 | 257 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15orf56| Gene Symbol: C15orf56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039905.2 | NP_001034994.1 | 1, 2 | 257 | ||
| GenBank mRNA | AK095915.1 | - | 1, 2 | 257 | ||