siRNA Details
Gene Symbol: MEIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080836.2 | NP_001074305.1 | 2 | 247 | ||
GenBank mRNA | AA971146.1 | - | 2 | 214 | ||
BG189536.1 | - | 1 | 131 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MEIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080836.2 | NP_001074305.1 | 2 | 247 | ||
GenBank mRNA | AA971146.1 | - | 2 | 214 | ||
BG189536.1 | - | 1 | 131 |