siRNA Details
| Gene Symbol: LRCOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195520.1 | NP_001182449.1 | 3, 4 | 378 | ||
| XM_011534780.2 | XP_011533082.1 | 3, 4 | 378 | |||
| XM_017018659.1 | XP_016874148.1 | 2, 3 | 354 | |||
| XM_017018660.1 | XP_016874149.1 | 2, 3 | 354 | |||
| GenBank mRNA | DB344052.1 | - | 3, 4 | 491 | ||
| JQ012741.1 | - | 3, 4 | 301 | |||
| JQ012742.1 | - | 3, 4 | 433 | |||

LRCOL1
Genomic Map