siRNA Details
| Gene Symbol: LINC01625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033919.1 | 3 | 174 | |||
| GenBank mRNA | AK123801.1 | - | 3 | 174 | ||
| BX108499.1 | - | 3 | 159 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01625| Gene Symbol: LINC01625 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033919.1 | 3 | 174 | |||
| GenBank mRNA | AK123801.1 | - | 3 | 174 | ||
| BX108499.1 | - | 3 | 159 | |||