siRNA Details
| Gene Symbol: C16orf90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080524.1 | NP_001073993.1 | 2 | 340 | ||
| XM_005255502.3 | XP_005255559.1 | 2 | 406 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16orf90| Gene Symbol: C16orf90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080524.1 | NP_001073993.1 | 2 | 340 | ||
| XM_005255502.3 | XP_005255559.1 | 2 | 406 | |||