siRNA Details
| Gene Symbol: ARIH2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123040.1 | NP_001116512.1 | 1 | 781 | ||
| GenBank mRNA | AK097724.1 | - | 1 | 781 | ||
| DA461567.1 | - | 1 | 371 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARIH2OS| Gene Symbol: ARIH2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123040.1 | NP_001116512.1 | 1 | 781 | ||
| GenBank mRNA | AK097724.1 | - | 1 | 781 | ||
| DA461567.1 | - | 1 | 371 | |||