siRNA Details
| Gene Symbol: C3orf84 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080528.2 | NP_001073997.2 | 4 | 591 | ||
| XM_011534015.2 | XP_011532317.1 | 4 | 1418 | |||
| XM_011534017.2 | XP_011532319.1 | 3 | 1306 | |||
| XM_011534018.2 | XP_011532320.1 | 4 | 1218 | |||

C3orf84
Genomic Map