siRNA Details
Gene Symbol: ZAR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136571.1 | NP_001130043.1 | 1 | 76 | ||
GenBank mRNA | BC146956.1 | - | 1 | 74 | 9056884 | |
BC146965.1 | - | 1 | 76 | 9056893 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZAR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136571.1 | NP_001130043.1 | 1 | 76 | ||
GenBank mRNA | BC146956.1 | - | 1 | 74 | 9056884 | |
BC146965.1 | - | 1 | 76 | 9056893 |