siRNA Details
| Gene Symbol: SH2D7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101404.1 | NP_001094874.1 | 1 | 150 | ||
| XM_011521895.2 | XP_011520197.1 | 1 | 1442 | |||
| XM_011521896.2 | XP_011520198.1 | 1 | 68 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH2D7| Gene Symbol: SH2D7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101404.1 | NP_001094874.1 | 1 | 150 | ||
| XM_011521895.2 | XP_011520197.1 | 1 | 1442 | |||
| XM_011521896.2 | XP_011520198.1 | 1 | 68 | |||