siRNA Details
Gene Symbol: GORASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031899.3 | NP_114105.1 | 2 | 932 | ||
NR_103866.1 | 2 | 469 | ||||
NR_103867.1 | 2 | 469 | ||||
XM_006713301.3 | XP_006713364.1 | 2 | 1032 | |||
XM_011534020.2 | XP_011532322.1 | 2 | 1032 | |||
XM_011534021.2 | XP_011532323.1 | 2 | 1032 | |||
XM_017007050.1 | XP_016862539.1 | 2 | 208 | |||
XM_017007051.1 | XP_016862540.1 | 2 | 647 | |||
XM_017007052.1 | XP_016862541.1 | 2 | 194 | |||
GenBank mRNA | AJ409349.1 | - | 2 | 208 | ||
AK001574.1 | - | 2 | 207 | |||
AK091168.1 | - | 2 | 396 | |||
AK124755.1 | - | 2 | 932 | |||
AK300271.1 | - | 2 | 148 | |||
BC008928.2 | - | 2 | 190 | |||
BC075854.1 | - | 2 | 218 | 5503263 | ||
BC103826.1 | - | 2 | 150 | 40001584 | ||
BC103827.1 | - | 2 | 150 | 40001588 | ||
DB031221.1 | - | 2 | 207 |
