siRNA Details
| Gene Symbol: FAM188B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | HM626137.1 | - | 5 | 549 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM188B2| Gene Symbol: FAM188B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | HM626137.1 | - | 5 | 549 | ||