siRNA Details
Gene Symbol: RD3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257268.1 | NP_001244197.1 | 3 | 761 | ||
GenBank mRNA | BQ184214.1 | - | 1 | 453 | ||
BQ185906.1 | - | 2 | 280 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RD3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257268.1 | NP_001244197.1 | 3 | 761 | ||
GenBank mRNA | BQ184214.1 | - | 1 | 453 | ||
BQ185906.1 | - | 2 | 280 |