siRNA Details
| Gene Symbol: RD3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257268.1 | NP_001244197.1 | 3 | 807 | ||
| GenBank mRNA | BQ184214.1 | - | 1 | 407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RD3L| Gene Symbol: RD3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257268.1 | NP_001244197.1 | 3 | 807 | ||
| GenBank mRNA | BQ184214.1 | - | 1 | 407 | ||