siRNA Details
| Gene Symbol: PLPPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022737.2 | NP_073574.2 | 4 | 519 | ||
| XM_005260017.1 | XP_005260074.1 | 4 | 519 | |||
| XM_017027147.1 | XP_016882636.1 | 4 | 522 | |||
| GenBank mRNA | AK023117.1 | - | 4 | 480 | ||
| AK225482.1 | - | 4 | 434 | |||
| AY304516.1 | - | 4 | 474 | |||
| AY339628.1 | - | 3 | 174 | |||
| BC009378.1 | - | 4 | 362 | 4125316 | ||

PLPPR2
Genomic Map