siRNA Details
Gene Symbol: CCDC136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022742.4 | NP_073579.4 | 11 | 2039 | ||
XM_005250534.3 | XP_005250591.1 | 12 | 1823 | |||
XM_005250538.1 | XP_005250595.1 | 12 | 1823 | |||
XM_011516483.1 | XP_011514785.1 | 12 | 1823 | |||
XM_011516484.1 | XP_011514786.1 | 12 | 1823 | |||
XM_011516485.1 | XP_011514787.1 | 12 | 1823 | |||
XM_011516486.1 | XP_011514788.1 | 12 | 1787 | |||
XM_011516487.2 | XP_011514789.1 | 12 | 1823 | |||
XM_011516488.2 | XP_011514790.1 | 12 | 1823 | |||
XM_011516489.1 | XP_011514791.1 | 12 | 1823 | |||
XM_011516490.2 | XP_011514792.1 | 12 | 1823 | |||
XM_017012532.1 | XP_016868021.1 | 12 | 1787 | |||
XM_017012533.1 | XP_016868022.1 | 12 | 2289 | |||
XM_017012534.1 | XP_016868023.1 | 12 | 1787 | |||
GenBank mRNA | AB058696.1 | - | 11 | 1990 | ||
AK023500.1 | - | 4 | 396 | |||
AL133027.1 | - | 8 | 1253 | |||
BC070114.1 | - | 7 | 1685 | |||
BC150331.1 | - | 11 | 2001 | 8860498 | ||
BC152408.1 | - | 11 | 1990 | 8862511 |
