siRNA Details
Gene Symbol: MUC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173600.2 | NP_775871.2 | 9074 | |||
GenBank mRNA | HM801842.1 | - | 9074 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MUC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173600.2 | NP_775871.2 | 9074 | |||
GenBank mRNA | HM801842.1 | - | 9074 |