siRNA Details
| Gene Symbol: C6orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022758.5 | NP_073595.2 | 1, 2 | 410 | ||
| NM_024294.3 | NP_077270.1 | 1, 2 | 410 | |||
| XM_005249298.2 | XP_005249355.1 | 1, 2 | 357 | |||
| GenBank mRNA | AK313410.1 | - | 1, 2 | 316 | ||
| BC002328.1 | - | 1, 2 | 324 | 3504258 | ||
| BC010184.1 | - | 1, 2 | 324 | 3959622 | ||
| BC075810.1 | - | 1, 2 | 332 | 5535440 | ||
| BE898102.1 | - | 1, 2 | 423 | |||

C6orf106
Genomic Map