siRNA Details
| Gene Symbol: CERK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022766.5 | NP_073603.2 | 9, 10 | 1162 | ||
| XM_017028909.1 | XP_016884398.1 | 9, 10 | 959 | |||
| GenBank mRNA | AB051433.1 | - | 8, 9 | 888 | ||
| AB079066.1 | - | 9, 10 | 1178 | |||
| AJ457828.1 | - | 9, 10 | 1125 | |||
| AK291476.1 | - | 9, 10 | 1118 | |||
| BC067255.1 | - | 8, 9 | 1039 | |||
| BC126940.1 | - | 9, 10 | 1105 | 40084110 | ||
| CR456404.1 | - | 9, 10 | 1065 | |||

CERK
Genomic Map