siRNA Details
| Gene Symbol: KLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134774.1 | NP_001128246.1 | 13 | 1620 | ||
| NM_001134775.1 | NP_001128247.1 | 14 | 1914 | |||
| NM_001134776.1 | NP_001128248.1 | 14 | 1793 | |||
| NM_001318734.1 | NP_001305663.1 | 14 | 1851 | |||
| NM_022822.2 | NP_073733.1 | 14 | 1888 | |||
| XM_005274183.1 | XP_005274240.1 | 14 | 1830 | |||
| XM_005274184.1 | XP_005274241.1 | 14 | 1851 | |||
| XM_017018133.1 | XP_016873622.1 | 12 | 1787 | |||
| GenBank mRNA | AK022449.1 | - | 14 | 1780 | ||
| AK022907.1 | - | 14 | 1833 | |||
| AK094593.1 | - | 12 | 1490 | |||
| AK126489.1 | - | 12 | 2271 | |||
| AK315725.1 | - | 14 | 1783 | |||
| AL136864.1 | - | 14 | 1814 | |||
| BC034373.1 | - | 14 | 1776 | 4342879 | ||

KLC2
Genomic Map