siRNA Details
| Gene Symbol: ACTG1P17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036446.1 | 3 | 744 | |||
| GenBank mRNA | BC020234.1 | - | 3 | 744 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACTG1P17| Gene Symbol: ACTG1P17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036446.1 | 3 | 744 | |||
| GenBank mRNA | BC020234.1 | - | 3 | 744 | ||