siRNA Details
| Gene Symbol: DLGAP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024101.1 | 3 | 517 | |||
| GenBank mRNA | AK093095.1 | - | 3 | 517 | ||
| BC137437.1 | - | 2 | 400 | |||
| BX647607.1 | - | 1 | 2460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLGAP1-AS1| Gene Symbol: DLGAP1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024101.1 | 3 | 517 | |||
| GenBank mRNA | AK093095.1 | - | 3 | 517 | ||
| BC137437.1 | - | 2 | 400 | |||
| BX647607.1 | - | 1 | 2460 | |||