siRNA Details
| Gene Symbol: INTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324475.1 | NP_001311404.1 | 31 | 3308 | ||
| NM_023015.4 | NP_075391.3 | 30 | 3649 | |||
| XM_005245459.2 | XP_005245516.1 | 29 | 3864 | |||
| XM_005245461.2 | XP_005245518.1 | 24 | 2339 | |||
| XM_006711490.3 | XP_006711553.1 | 30 | 3317 | |||
| XM_011509906.2 | XP_011508208.1 | 29 | 3229 | |||
| XM_017002132.1 | XP_016857621.1 | 19 | 2105 | |||
| GenBank mRNA | AF130066.2 | - | 1 | 947 | ||
| AK025572.1 | - | 16 | 1470 | |||
| AK055174.1 | - | 9 | 2032 | |||
| AK074979.1 | - | 23 | 2347 | |||
| AK094402.1 | - | 1 | 581 | |||
| AK126799.1 | - | 10 | 2187 | |||
| AK225294.1 | - | 16 | 1470 | |||
| AK290025.1 | - | 31 | 3302 | |||
| AK298746.1 | - | 18 | 1799 | |||
| AK304874.1 | - | 23 | 2936 | |||
| AL080172.1 | - | 25 | 2548 | |||
| BC025254.1 | - | 14 | 1309 | |||
| BC054513.1 | - | 17 | 1685 | |||
| BC073985.1 | - | 14 | 1321 | |||
| BC098431.1 | - | 16 | 1509 | |||
| BC105092.1 | - | 30 | 3112 | 8144095 | ||
| BC105094.1 | - | 30 | 3112 | 8144097 | ||
| BC116458.1 | - | 30 | 3644 | 5752962 | ||
| BX640786.1 | - | 30 | 3635 | |||
| BX640950.1 | - | 30 | 3650 | |||
| CR749212.1 | - | 28 | 5176 | |||
| CR749376.1 | - | 29 | 5301 | |||

INTS3
Genomic Map