siRNA Details
| Gene Symbol: FLJ38576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046264.1 | 1 | 666 | |||
| GenBank mRNA | AK095895.1 | - | 1 | 669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FLJ38576| Gene Symbol: FLJ38576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046264.1 | 1 | 666 | |||
| GenBank mRNA | AK095895.1 | - | 1 | 669 | ||