siRNA Details
| Gene Symbol: SLC2A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042.2 | NP_001033.1 | 5 | 729 | ||
| GenBank mRNA | AK300899.1 | - | 5 | 796 | ||
| AK313372.1 | - | 5 | 743 | |||
| AL521328.3 | - | 5 | 728 | |||
| BC034387.1 | - | 5 | 729 | 5187454 | ||
| BC069615.1 | - | 5 | 624 | 7262457 | ||
| BC069621.1 | - | 5 | 624 | 7262469 | ||
| BC113592.1 | - | 5 | 556 | 8322644 | ||
| BC126164.1 | - | 5 | 556 | 8991880 | ||
| M20747.1 | - | 5 | 674 | |||

SLC2A4
Genomic Map