siRNA Details
| Gene Symbol: WI2-2373I1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195127.1 | NP_001182056.1 | 1, 2 | 108 | ||
| XM_011515056.1 | XP_011513358.1 | 1, 2 | 108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WI2-2373I1.2| Gene Symbol: WI2-2373I1.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195127.1 | NP_001182056.1 | 1, 2 | 108 | ||
| XM_011515056.1 | XP_011513358.1 | 1, 2 | 108 | |||