siRNA Details
| Gene Symbol: MZT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005263741.4 | XP_005263798.1 | 3 | 674 | ||
| GenBank mRNA | AK090531.1 | - | 1 | 1213 | ||
| Gene Symbol: MZT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011511886.2 | XP_011510188.1 | 3 | 430 | ||
| XM_011511887.2 | XP_011510189.1 | 3 | 397 | |||
| XM_011511888.2 | XP_011510190.1 | 3 | 731 | |||
| GenBank mRNA | AK024408.1 | - | 1 | 588 | ||

MZT2A
Genomic Map