siRNA Details
| Gene Symbol: MZT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005263741.4 | XP_005263798.1 | 3 | 744 | ||
| GenBank mRNA | AK090531.1 | - | 1 | 1283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MZT2A| Gene Symbol: MZT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005263741.4 | XP_005263798.1 | 3 | 744 | ||
| GenBank mRNA | AK090531.1 | - | 1 | 1283 | ||