siRNA Details
| Gene Symbol: SLC12A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000338.2 | NP_000329.2 | 8 | 1219 | ||
| NM_001184832.1 | NP_001171761.1 | 8 | 1219 | |||
| XM_005254606.1 | XP_005254663.1 | 8 | 1219 | |||
| GenBank mRNA | AK295375.1 | - | 7 | 672 | ||
| AK298275.1 | - | 3 | 438 | |||
| AK298312.1 | - | 8 | 1218 | |||
| BC040138.2 | - | 7 | 1189 | |||
| BX647067.1 | - | 3 | 5142 | |||
| BX647484.1 | - | 4 | 5126 | |||
| EF559316.2 | - | 7 | 1017 | |||
| U58130.1 | - | 7 | 1022 | |||

SLC12A1
Genomic Map