siRNA Details
| Gene Symbol: BMPR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001203.2 | NP_001194.1 | 10 | 1234 | ||
| NM_001256792.1 | NP_001243721.1 | 8 | 1130 | |||
| NM_001256793.1 | NP_001243722.1 | 8 | 1071 | |||
| NM_001256794.1 | NP_001243723.1 | 8 | 1114 | |||
| XM_011532201.2 | XP_011530503.1 | 9 | 1113 | |||
| XM_017008558.1 | XP_016864047.1 | 10 | 1305 | |||
| XM_017008559.1 | XP_016864048.1 | 11 | 1295 | |||
| XM_017008560.1 | XP_016864049.1 | 8 | 1187 | |||
| XM_017008561.1 | XP_016864050.1 | 9 | 1144 | |||
| GenBank mRNA | AK293008.1 | - | 8 | 1114 | ||
| AK299930.1 | - | 8 | 1071 | |||
| AK313642.1 | - | 8 | 1130 | |||
| BC047773.1 | - | 7 | 1030 | 6202951 | ||
| BC069796.1 | - | 9 | 1146 | 7262484 | ||
| BC069803.1 | - | 9 | 1146 | 7262496 | ||
| D89675.1 | - | 7 | 978 | |||
| U89326.1 | - | 10 | 1233 | |||

BMPR1B
Genomic Map